Assay Details
| Gene Symbol: Xylb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033209.3 | NP_001028381.1 | 11 - 12 | 1157 | 86 | |
| NM_001199568.1 | NP_001186497.1 | 12 - 13 | 1205 | 86 | ||
| GenBank mRNA | AK143476.1 | - | 11 - 12 | 1157 | 86 | |
| AK164186.1 | - | 12 - 13 | 999 | 86 | ||
| AK165440.1 | - | 11 - 12 | 954 | 86 | ||
| AK165518.1 | - | 11 - 12 | 954 | 86 | ||
| AK165759.1 | - | 12 - 13 | 1001 | 86 | ||
| AK167735.1 | - | 2 - 3 | 177 | 86 | ||
| AK170197.1 | - | 10 - 11 | 902 | 86 | ||
| BC025442.1 | - | 11 - 12 | 907 | 86 | ||
| BC080743.1 | - | 11 - 12 | 931 | 86 | ||
| BC138244.1 | - | 11 - 12 | 1106 | 8861263 | 86 | |
| BC138247.1 | - | 11 - 12 | 1106 | 8861266 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xylb
Genomic Map