Assay Details
| Gene Symbol: Xylb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033209.3 | NP_001028381.1 | 16 - 17 | 1616 | 88 | |
| NM_001199568.1 | NP_001186497.1 | 17 - 18 | 1664 | 88 | ||
| GenBank mRNA | AK143476.1 | - | 16 - 17 | 1616 | 88 | |
| AK164186.1 | - | 17 - 18 | 1458 | 88 | ||
| AK165440.1 | - | 16 - 17 | 1413 | 88 | ||
| AK165518.1 | - | 16 - 17 | 1413 | 88 | ||
| AK165759.1 | - | 17 - 18 | 1460 | 88 | ||
| AK167735.1 | - | 7 - 8 | 636 | 88 | ||
| AK170197.1 | - | 15 - 16 | 1361 | 88 | ||
| BC025442.1 | - | 16 - 17 | 1366 | 88 | ||
| BC026406.1 | - | 4 - 5 | 328 | 88 | ||
| BC080743.1 | - | 16 - 17 | 1390 | 88 | ||
| BC138244.1 | - | 16 - 17 | 1565 | 8861263 | 88 | |
| BC138247.1 | - | 16 - 17 | 1565 | 8861266 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xylb
Genomic Map