Assay Details
| Gene Symbol: Xylb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033209.3 | NP_001028381.1 | 7 - 8 | 844 | 67 | |
| GenBank mRNA | AK143476.1 | - | 7 - 8 | 844 | 67 | |
| AK165440.1 | - | 7 - 8 | 641 | 67 | ||
| AK165518.1 | - | 7 - 8 | 641 | 67 | ||
| AK180117.1 | - | 2 - 3 | 126 | 67 | ||
| BC025442.1 | - | 7 - 8 | 594 | 67 | ||
| BC080743.1 | - | 7 - 8 | 618 | 67 | ||
| BC138244.1 | - | 7 - 8 | 793 | 8861263 | 67 | |
| BC138247.1 | - | 7 - 8 | 793 | 8861266 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xylb
Genomic Map