Assay Details
| Gene Symbol: Xylb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033209.3 | NP_001028381.1 | 9 - 10 | 1032 | 80 | |
| NM_001199568.1 | NP_001186497.1 | 10 - 11 | 1080 | 80 | ||
| GenBank mRNA | AK143476.1 | - | 9 - 10 | 1032 | 80 | |
| AK164186.1 | - | 10 - 11 | 874 | 80 | ||
| AK165440.1 | - | 9 - 10 | 829 | 80 | ||
| AK165518.1 | - | 9 - 10 | 829 | 80 | ||
| AK165759.1 | - | 10 - 11 | 876 | 80 | ||
| AK170197.1 | - | 8 - 9 | 777 | 80 | ||
| BC025442.1 | - | 9 - 10 | 782 | 80 | ||
| BC080743.1 | - | 9 - 10 | 806 | 80 | ||
| BC138244.1 | - | 9 - 10 | 981 | 8861263 | 80 | |
| BC138247.1 | - | 9 - 10 | 981 | 8861266 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xylb
Genomic Map