Assay Details
| Gene Symbol: Pik3r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008841.2 | NP_032867.2 | 11 - 12 | 1879 | 69 | |
| GenBank mRNA | AK146195.1 | - | 11 - 12 | 1869 | 69 | |
| AK148122.1 | - | 11 - 12 | 1876 | 69 | ||
| BC006796.1 | - | 11 - 12 | 1870 | 2581969 | 69 | |
| BC085501.1 | - | 10 - 11 | 1802 | 6816009 | 69 | |
| Y13569.1 | - | 11 - 12 | 1848 | 69 | ||
More Information
Set Membership: |
|

Pik3r2
Genomic Map