Assay Details
| Gene Symbol: Pik3r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008841.2 | NP_032867.2 | 14 - 15 | 2273 | 61 | |
| GenBank mRNA | AK148122.1 | - | 14 - 15 | 2270 | 61 | |
| BC006796.1 | - | 14 - 15 | 2264 | 2581969 | 61 | |
| BC085501.1 | - | 13 - 14 | 2196 | 6816009 | 61 | |
| Y13569.1 | - | 14 - 15 | 2242 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pik3r2
Genomic Map