Assay Details
| Gene Symbol: Pik3r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008841.2 | NP_032867.2 | 6 - 7 | 1275 | 70 | |
| GenBank mRNA | AK146195.1 | - | 6 - 7 | 1265 | 70 | |
| AK148122.1 | - | 6 - 7 | 1272 | 70 | ||
| BC006796.1 | - | 6 - 7 | 1266 | 2581969 | 70 | |
| BC085501.1 | - | 5 - 6 | 1198 | 6816009 | 70 | |
| Y13569.1 | - | 6 - 7 | 1244 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pik3r2
Genomic Map