Assay Details
| Gene Symbol: Pik3r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008841.2 | NP_032867.2 | 9 - 10 | 1571 | 74 | |
| GenBank mRNA | AK146195.1 | - | 9 - 10 | 1561 | 74 | |
| AK148122.1 | - | 9 - 10 | 1568 | 74 | ||
| BC006796.1 | - | 9 - 10 | 1562 | 2581969 | 74 | |
| BC085501.1 | - | 8 - 9 | 1494 | 6816009 | 74 | |
| Y13569.1 | - | 9 - 10 | 1540 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pik3r2
Genomic Map