Assay Details
| Gene Symbol: Kcnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF357239.1 | - | 7 - 8 | 1406 | 70 | |
| AK041033.1 | - | 7 - 8 | 1451 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnn1| Gene Symbol: Kcnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF357239.1 | - | 7 - 8 | 1406 | 70 | |
| AK041033.1 | - | 7 - 8 | 1451 | 70 | ||
Set Membership: |
|