Assay Details
| Gene Symbol: Slc5a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF380353.1 | - | 11 - 12 | 1345 | 86 | |
| BC137650.1 | - | 11 - 12 | 1358 | 8860669 | 86 | |
| BC137651.1 | - | 11 - 12 | 1358 | 8860670 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc5a5
Genomic Map