Assay Details
| Gene Symbol: Aadat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011834.2 | NP_035964.1 | 2 - 3 | 326 | 113 | |
| GenBank mRNA | AF072376.1 | - | 2 - 3 | 326 | 113 | |
| AK075578.1 | - | 2 - 3 | 332 | 113 | ||
| AK144042.1 | - | 2 - 3 | 279 | 113 | ||
| BC012637.1 | - | 2 - 3 | 329 | 4160508 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aadat
Genomic Map