Assay Details
| Gene Symbol: Aadat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011834.2 | NP_035964.1 | 6 - 7 | 803 | 156 | |
| GenBank mRNA | AF072376.1 | - | 6 - 7 | 803 | 156 | |
| AK075578.1 | - | 6 - 7 | 809 | 156 | ||
| AK144042.1 | - | 6 - 7 | 756 | 156 | ||
| BC012637.1 | - | 6 - 7 | 806 | 4160508 | 156 | |
More Information
Set Membership: |
|

Aadat
Genomic Map