| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300011396.mm.1 |
NM_025965.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011396.mm.1 |
NM_025965.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011396.mm.1 |
TC1300001827.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300011398.mm.1 |
NM_025965.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011398.mm.1 |
NM_025965.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011398.mm.1 |
TC1300001827.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300011394.mm.1 |
NM_025965.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011394.mm.1 |
NM_025965.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011394.mm.1 |
TC1300001827.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300011397.mm.1 |
NM_025965.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011397.mm.1 |
NM_025965.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011397.mm.1 |
TC1300001827.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300011395.mm.1 |
NM_025965.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011395.mm.1 |
NM_025965.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011395.mm.1 |
TC1300001827.mm.1 |
MTA TC |