Assay Details
| Gene Symbol: Ssr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025965.3 | NP_080241.3 | 6 - 7 | 801 | 106 | |
| GenBank mRNA | AF326229.2 | - | 6 - 7 | 781 | 106 | |
| AF395811.1 | - | 6 - 7 | 781 | 106 | ||
| AK010884.1 | - | 6 - 7 | 747 | 106 | ||
| AK031538.1 | - | 6 - 7 | 781 | 106 | ||
| AK160285.1 | - | 6 - 7 | 764 | 106 | ||
| AK166235.1 | - | 6 - 7 | 775 | 106 | ||
| AK167793.1 | - | 6 - 7 | 747 | 106 | ||
| AY993942.1 | - | 781 | 106 | |||
| BC011255.1 | - | 6 - 7 | 736 | 4017600 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ssr1
Genomic Map