Assay Details
| Gene Symbol: Nup153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175749.2 | NP_786925.2 | 11 - 12 | 1770 | 104 | |
| GenBank mRNA | BC052173.1 | - | 11 - 12 | 1508 | 6516328 | 104 |
| CA326562.1 | - | 6 - 7 | 462 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nup153
Genomic Map