Assay Details
| Gene Symbol: Nup153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175749.2 | NP_786925.2 | 16 - 17 | 2816 | 126 | |
| GenBank mRNA | BC046468.1 | - | 4 - 5 | 840 | 126 | |
| BC052173.1 | - | 16 - 17 | 2554 | 6516328 | 126 | |
| CF897724.1 | - | 1 - 2 | 582 | 126 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nup153
Genomic Map