Assay Details
| Gene Symbol: Txn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011660.3 | NP_035790.1 | 4 - 5 | 457 | 126 | |
| GenBank mRNA | AK007537.1 | - | 4 - 5 | 338 | 126 | |
| AK007790.1 | - | 4 - 5 | 335 | 126 | ||
| BC010756.1 | - | 4 - 5 | 312 | 4216106 | 126 | |
| BC094415.1 | - | 4 - 5 | 319 | 3376881 | 126 | |
| X77585.1 | - | 4 - 5 | 330 | 126 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Txn1
Genomic Map