Assay Details
| Gene Symbol: Azi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048146.2 | NP_001041611.1 | 2 - 3 | 656 | 141 | |
| NM_001286507.1 | NP_001273436.1 | 2 - 3 | 656 | 141 | ||
| NM_001286508.1 | NP_001273437.1 | 2 - 3 | 574 | 141 | ||
| NM_013727.4 | NP_038755.1 | 2 - 3 | 656 | 141 | ||
| GenBank mRNA | AB007141.1 | - | 2 - 3 | 492 | 141 | |
| AK004992.1 | - | 2 - 3 | 635 | 141 | ||
| AK015942.1 | - | 2 - 3 | 477 | 141 | ||
| AK030076.1 | - | 2 - 3 | 488 | 141 | ||
| AK036102.1 | - | 2 - 3 | 563 | 141 | ||
| AK134714.1 | - | 2 - 3 | 559 | 141 | ||
| AK153477.1 | - | 2 - 3 | 620 | 141 | ||
| AK165549.1 | - | 2 - 3 | 542 | 141 | ||
| AK167783.1 | - | 2 - 3 | 600 | 141 | ||
| BC006851.1 | - | 2 - 3 | 551 | 3156857 | 141 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001286507.1 transcripts.
Set Membership: |
|

Azi2
Genomic Map