Assay Details
| Gene Symbol: Entpd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178676.4 | NP_848791.2 | 6 - 7 | 675 | 106 | |
| GenBank mRNA | AK032057.1 | - | 6 - 7 | 661 | 106 | |
| AK046218.1 | - | 6 - 7 | 651 | 106 | ||
| AK087561.1 | - | 6 - 7 | 649 | 106 | ||
| AK169947.1 | - | 1 - 2 | 85 | 106 | ||
| AK170974.1 | - | 1 - 2 | 84 | 106 | ||
| AY376710.1 | - | 6 - 7 | 643 | 106 | ||
| AY714060.1 | - | 6 - 7 | 646 | 106 | ||
| BC079871.1 | - | 6 - 7 | 646 | 6831925 | 106 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK032057.1 transcripts.
Set Membership: |
|

Entpd3
Genomic Map