Assay Details
| Gene Symbol: Tll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009390.2 | NP_033416.2 | 2 - 3 | 885 | 66 | |
| GenBank mRNA | AK080919.1 | - | 2 - 3 | 901 | 66 | |
| U34042.1 | - | 2 - 3 | 885 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tll1
Genomic Map