Assay Details
| Gene Symbol: Tll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009390.2 | NP_033416.2 | 3 - 4 | 970 | 106 | |
| GenBank mRNA | AK080919.1 | - | 3 - 4 | 986 | 106 | |
| U34042.1 | - | 3 - 4 | 970 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tll1
Genomic Map