Assay Details
| Gene Symbol: Larp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028451.1 | NP_082727.1 | 7 - 8 | 1292 | 84 | |
| GenBank mRNA | AK138840.1 | - | 4 - 5 | 638 | 84 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Larp1
Genomic Map