Assay Details
| Gene Symbol: Larp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028451.1 | NP_082727.1 | 8 - 9 | 1431 | 61 | |
| GenBank mRNA | AK138840.1 | - | 5 - 6 | 777 | 61 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Larp1
Genomic Map