Assay Details
| Gene Symbol: Tert | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009354.1 | NP_033380.1 | 11 - 12 | 2848 | 78 | |
| GenBank mRNA | AF051911.1 | - | 11 - 12 | 2848 | 78 | |
| AF073311.1 | - | 11 - 12 | 2819 | 78 | ||
| BC082327.1 | - | 12 - 13 | 2350 | 78 | ||
| BC127068.1 | - | 12 - 13 | 2136 | 40110173 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tert
Genomic Map