Assay Details
| Gene Symbol: Tert | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009354.1 | NP_033380.1 | 8 - 9 | 2471 | 91 | |
| GenBank mRNA | AF051911.1 | - | 8 - 9 | 2471 | 91 | |
| AF073311.1 | - | 8 - 9 | 2442 | 91 | ||
| BC082327.1 | - | 9 - 10 | 1973 | 91 | ||
| BC127068.1 | - | 9 - 10 | 1759 | 40110173 | 91 | |
| FJ210936.1 | - | 4 - 5 | 467 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tert
Genomic Map