Assay Details
| Gene Symbol: Slc12a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011390.2 | NP_035520.1 | 11 - 12 | 1528 | 66 | |
| GenBank mRNA | AF087436.1 | - | 11 - 12 | 1530 | 66 | |
| AK131129.1 | - | 11 - 12 | 1482 | 66 | ||
| AK143535.1 | - | 11 - 12 | 1522 | 66 | ||
| AK149750.1 | - | 11 - 12 | 1523 | 66 | ||
| AK166215.1 | - | 11 - 12 | 1522 | 66 | ||
| AK169950.1 | - | 11 - 12 | 1518 | 66 | ||
| AK171498.1 | - | 11 - 12 | 1565 | 66 | ||
| BC059242.1 | - | 11 - 12 | 1520 | 66 | ||
| BC060133.1 | - | 11 - 12 | 1520 | 66 | ||
| BC141107.1 | - | 11 - 12 | 1488 | 9055719 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc12a7
Genomic Map