Assay Details
| Gene Symbol: Slc12a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011390.2 | NP_035520.1 | 12 - 13 | 1694 | 57 | |
| GenBank mRNA | AF087436.1 | - | 12 - 13 | 1696 | 57 | |
| AK131129.1 | - | 12 - 13 | 1648 | 57 | ||
| AK143535.1 | - | 12 - 13 | 1688 | 57 | ||
| AK149750.1 | - | 12 - 13 | 1689 | 57 | ||
| AK169950.1 | - | 12 - 13 | 1684 | 57 | ||
| AK171498.1 | - | 12 - 13 | 1731 | 57 | ||
| BC059242.1 | - | 12 - 13 | 1686 | 57 | ||
| BC060133.1 | - | 12 - 13 | 1686 | 57 | ||
| BC079677.1 | - | 11 - 12 | 2354 | 57 | ||
| BC141107.1 | - | 12 - 13 | 1654 | 9055719 | 57 | |
More Information
Set Membership: |
|

Slc12a7
Genomic Map