Assay Details
| Gene Symbol: Slc12a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011390.2 | NP_035520.1 | 18 - 19 | 2505 | 78 | |
| GenBank mRNA | AF087436.1 | - | 18 - 19 | 2507 | 78 | |
| AK131129.1 | - | 18 - 19 | 2459 | 78 | ||
| AK143535.1 | - | 18 - 19 | 2499 | 78 | ||
| AK149750.1 | - | 18 - 19 | 2500 | 78 | ||
| AK164405.1 | - | 1 - 2 | 999 | 78 | ||
| AK169950.1 | - | 18 - 19 | 2495 | 78 | ||
| AK171498.1 | - | 18 - 19 | 2542 | 78 | ||
| BC079677.1 | - | 15 - 16 | 4255 | 78 | ||
| BC141107.1 | - | 18 - 19 | 2465 | 9055719 | 78 | |
More Information
Set Membership: |
|

Slc12a7
Genomic Map