Assay Details
| Gene Symbol: 2210407C18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144544.2 | NP_653127.2 | 4 - 5 | 443 | 74 | |
| GenBank mRNA | AA124355.1 | - | 4 - 5 | 429 | 74 | |
| AA434919.1 | - | 2 - 3 | 249 | 74 | ||
| AY148487.1 | - | 3 - 4 | 349 | 74 | ||
| BC019553.1 | - | 4 - 5 | 421 | 4456658 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1992546 | BC019553.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 29326 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235327 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 162301 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 784496 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 22368 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147313 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1992546 | BC019553.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831880 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147313 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07357759 | - | - |
| Whole Mouse Genome G4122A | A_51_P316673 | BC019553.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119955 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147313 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09469818 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09731483 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09469823 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09731489 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09731482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08981968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09052796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08981972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09698182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09702000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09469818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831901 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09050938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09450710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09702003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09469819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09053406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09266553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09184583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09731487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831906 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09702004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831880 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09009647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831874 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10386201 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10386200 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10386198 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10386204 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10386203 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10386199 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10386202 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425233_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10386197 | - | - |
| Mouse Genome 430 2.0 Array | 1425233_at | - | - |
| Mouse Genome 430 2.0 Array | 1425233_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78354_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78354_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100025569.mm.1 | NM_144544.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100025569.mm.1 | NM_144544.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100025569.mm.1 | TC1100002829.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425233_at | - | - |
| Mouse Expression Set 430 | 1425233_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425233_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425233_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425233_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425233_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6450037 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006450037 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1226767 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226767 | - | - |
| mouse-6 v1.1 (Illumina) | scl40181.5.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6450037 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40181.5.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6450037 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1226767 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226767 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl40181.5.1_1-S | - | - |

2210407C18Rik
Genomic Map