Assay Details
| Gene Symbol: Ano10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271873.1 | NP_001258802.1 | 10 - 11 | 1629 | 65 | |
| NM_133979.3 | NP_598740.1 | 10 - 11 | 1803 | 65 | ||
| GenBank mRNA | AK028391.1 | - | 5 - 6 | 748 | 65 | |
| AK043011.1 | - | 10 - 11 | 1691 | 65 | ||
| AK046091.1 | - | 10 - 11 | 1755 | 65 | ||
| AK054163.1 | - | 6 - 7 | 1093 | 65 | ||
| AK079622.1 | - | 10 - 11 | 1754 | 65 | ||
| AK080947.1 | - | 10 - 11 | 1755 | 65 | ||
| AK138414.1 | - | 10 - 11 | 1878 | 65 | ||
| AK186643.1 | - | 1 - 2 | 123 | 65 | ||
| BC002294.1 | - | 1 - 2 | 205 | 65 | ||
| BC026421.1 | - | 5 - 6 | 843 | 65 | ||
| BC099688.1 | - | 10 - 11 | 1552 | 6313494 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ano10
Genomic Map