Assay Details
| Gene Symbol: Ano10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133979.3 | NP_598740.1 | 5 - 6 | 725 | 64 | |
| GenBank mRNA | AK043011.1 | - | 5 - 6 | 688 | 64 | |
| AK046091.1 | - | 5 - 6 | 677 | 64 | ||
| AK079622.1 | - | 5 - 6 | 676 | 64 | ||
| AK080947.1 | - | 5 - 6 | 677 | 64 | ||
| AK138414.1 | - | 5 - 6 | 800 | 64 | ||
| AK194201.1 | - | 1 - 2 | 38 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ano10
Genomic Map