Assay Details
| Gene Symbol: Ano10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271873.1 | NP_001258802.1 | 8 - 9 | 1253 | 82 | |
| NM_133979.3 | NP_598740.1 | 8 - 9 | 1427 | 82 | ||
| GenBank mRNA | AK028391.1 | - | 3 - 4 | 372 | 82 | |
| AK043011.1 | - | 8 - 9 | 1390 | 82 | ||
| AK046091.1 | - | 8 - 9 | 1379 | 82 | ||
| AK054163.1 | - | 4 - 5 | 717 | 82 | ||
| AK079622.1 | - | 8 - 9 | 1378 | 82 | ||
| AK080947.1 | - | 8 - 9 | 1379 | 82 | ||
| AK138414.1 | - | 8 - 9 | 1502 | 82 | ||
| BC026421.1 | - | 3 - 4 | 467 | 82 | ||
| BC099688.1 | - | 8 - 9 | 1176 | 6313494 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ano10
Genomic Map