Assay Details
| Gene Symbol: Ncan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007789.3 | NP_031815.2 | 9 - 10 | 3117 | 72 | |
| GenBank mRNA | BC065118.1 | - | 9 - 10 | 3083 | 6853253 | 72 |
| X84727.1 | - | 9 - 10 | 3079 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ncan
Genomic Map