Assay Details
| Gene Symbol: Tm6sf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293795.1 | NP_001280724.1 | 5 - 6 | 550 | 85 | |
| NM_181540.4 | NP_853518.1 | 5 - 6 | 550 | 85 | ||
| GenBank mRNA | AK077126.1 | - | 5 - 6 | 543 | 85 | |
| BC024498.1 | - | 5 - 6 | 538 | 4982620 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tm6sf2
Genomic Map