Assay Details
| Gene Symbol: Isyna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023627.1 | NP_076116.1 | 2 - 3 | 199 | 87 | |
| GenBank mRNA | AF288525.1 | - | 2 - 3 | 199 | 87 | |
| AK005029.2 | - | 2 - 3 | 177 | 87 | ||
| AK079323.1 | - | 2 - 3 | 176 | 87 | ||
| BC003458.1 | - | 2 - 3 | 190 | 2648362 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Isyna1
Genomic Map