Assay Details
| Gene Symbol: Isyna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023627.1 | NP_076116.1 | 8 - 9 | 1214 | 93 | |
| GenBank mRNA | AF288525.1 | - | 8 - 9 | 1214 | 93 | |
| AK005029.2 | - | 8 - 9 | 1192 | 93 | ||
| AK079323.1 | - | 8 - 9 | 1191 | 93 | ||
| AK137089.1 | - | 7 - 8 | 1296 | 93 | ||
| BC003458.1 | - | 8 - 9 | 1205 | 2648362 | 93 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Isyna1
Genomic Map