Assay Details
| Gene Symbol: Zfp445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173364.5 | NP_775540.4 | 6 - 7 | 1114 | 154 | |
| GenBank mRNA | AY341877.1 | - | 5 - 6 | 955 | 154 | |
| BC099896.1 | - | 5 - 6 | 926 | 30545532 | 154 | |
| BC099930.1 | - | 6 - 7 | 1059 | 30058096 | 154 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp445
Genomic Map