Assay Details
| Gene Symbol: Tcaim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013405.2 | NP_001013423.2 | 1 - 2 | 140 | 82 | |
| GenBank mRNA | BC080690.1 | - | 1 - 2 | 99 | 30111134 | 82 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC080690.1, NM_001013405.2 transcripts.
Set Membership: |
|

Tcaim
Genomic Map