Assay Details
| Gene Symbol: Tmem161b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175187.5 | NP_780396.2 | 6 - 7 | 740 | 69 | |
| NR_029418.1 | 5 - 6 | 598 | 69 | |||
| NR_029419.1 | 6 - 7 | 740 | 69 | |||
| GenBank mRNA | AK013305.1 | - | 5 - 6 | 569 | 69 | |
| AK088393.1 | - | 6 - 7 | 712 | 69 | ||
| BC037611.1 | - | 4 - 5 | 520 | 69 | ||
| BC051663.1 | - | 6 - 7 | 688 | 30020101 | 69 | |
| BC060067.1 | - | 6 - 7 | 722 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem161b
Genomic Map