Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300006346.mm.1 |
NM_175187.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006346.mm.1 |
NM_175187.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006346.mm.1 |
NR_029418.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006346.mm.1 |
NR_029418.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006346.mm.1 |
NR_029419.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006346.mm.1 |
NR_029419.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006346.mm.1 |
TC1300000983.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006347.mm.1 |
NR_029419.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006347.mm.1 |
NR_029419.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006347.mm.1 |
TC1300000983.mm.1 |
MTA TC |