Assay Details
| Gene Symbol: Sdha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023281.1 | NP_075770.1 | 12 - 13 | 1697 | 106 | |
| GenBank mRNA | AF095938.1 | - | 10 - 11 | 1446 | 106 | |
| AK004362.1 | - | 2 - 3 | 248 | 106 | ||
| AK010561.1 | - | 12 - 13 | 1687 | 107 | ||
| AK029520.1 | - | 12 - 13 | 1697 | 106 | ||
| AK034928.1 | - | 12 - 13 | 1698 | 106 | ||
| AK049590.1 | - | 12 - 13 | 1684 | 106 | ||
| AK050475.1 | - | 12 - 13 | 1684 | 106 | ||
| AK075990.1 | - | 12 - 13 | 1681 | 106 | ||
| AK145923.1 | - | 12 - 13 | 1695 | 106 | ||
| AK147286.1 | - | 12 - 13 | 1684 | 106 | ||
| AK147624.1 | - | 12 - 13 | 1684 | 106 | ||
| AK151333.1 | - | 4 - 5 | 325 | 106 | ||
| AK153085.1 | - | 12 - 13 | 1704 | 106 | ||
| AK162148.1 | - | 12 - 13 | 1680 | 106 | ||
| AK169254.1 | - | 12 - 13 | 1680 | 106 | ||
| BC011301.1 | - | 12 - 13 | 1657 | 106 | ||
| DQ402975.1 | - | 10 - 11 | 1491 | 106 | ||
More Information
Set Membership: |
|

Sdha
Genomic Map