Assay Details
| Gene Symbol: Sdha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023281.1 | NP_075770.1 | 3 - 4 | 342 | 64 | |
| GenBank mRNA | AF095938.1 | - | 1 - 2 | 91 | 64 | |
| AK010561.1 | - | 3 - 4 | 331 | 64 | ||
| AK029520.1 | - | 3 - 4 | 342 | 64 | ||
| AK034928.1 | - | 3 - 4 | 343 | 64 | ||
| AK049590.1 | - | 3 - 4 | 329 | 64 | ||
| AK050475.1 | - | 3 - 4 | 329 | 64 | ||
| AK075990.1 | - | 3 - 4 | 326 | 64 | ||
| AK145923.1 | - | 3 - 4 | 340 | 64 | ||
| AK147286.1 | - | 3 - 4 | 329 | 64 | ||
| AK147624.1 | - | 3 - 4 | 329 | 64 | ||
| AK153085.1 | - | 3 - 4 | 349 | 64 | ||
| AK162148.1 | - | 3 - 4 | 325 | 64 | ||
| AK169254.1 | - | 3 - 4 | 325 | 64 | ||
| AK207923.1 | - | 2 - 3 | 218 | 64 | ||
| BC011301.1 | - | 3 - 4 | 302 | 64 | ||
| BC031849.1 | - | 3 - 4 | 350 | 5067510 | 64 | |
| DQ402975.1 | - | 1 - 2 | 136 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sdha
Genomic Map