Assay Details
| Gene Symbol: Sdha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023281.1 | NP_075770.1 | 5 - 6 | 654 | 90 | |
| GenBank mRNA | AK010561.1 | - | 5 - 6 | 643 | 90 | |
| AK029520.1 | - | 5 - 6 | 654 | 90 | ||
| AK034928.1 | - | 5 - 6 | 655 | 90 | ||
| AK049590.1 | - | 5 - 6 | 641 | 90 | ||
| AK050475.1 | - | 5 - 6 | 641 | 90 | ||
| AK075990.1 | - | 5 - 6 | 638 | 90 | ||
| AK145923.1 | - | 5 - 6 | 652 | 90 | ||
| AK147286.1 | - | 5 - 6 | 641 | 90 | ||
| AK147624.1 | - | 5 - 6 | 641 | 90 | ||
| AK153085.1 | - | 5 - 6 | 661 | 90 | ||
| AK162148.1 | - | 5 - 6 | 637 | 90 | ||
| AK169254.1 | - | 5 - 6 | 637 | 90 | ||
| BC031849.1 | - | 5 - 6 | 662 | 5067510 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sdha
Genomic Map