Assay Details
| Gene Symbol: Sdha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023281.1 | NP_075770.1 | 7 - 8 | 929 | 81 | |
| GenBank mRNA | AF095938.1 | - | 5 - 6 | 678 | 81 | |
| AK010561.1 | - | 7 - 8 | 918 | 81 | ||
| AK029520.1 | - | 7 - 8 | 929 | 81 | ||
| AK034928.1 | - | 7 - 8 | 930 | 81 | ||
| AK049590.1 | - | 7 - 8 | 916 | 81 | ||
| AK050475.1 | - | 7 - 8 | 916 | 81 | ||
| AK075990.1 | - | 7 - 8 | 913 | 81 | ||
| AK145923.1 | - | 7 - 8 | 927 | 81 | ||
| AK147286.1 | - | 7 - 8 | 916 | 81 | ||
| AK147624.1 | - | 7 - 8 | 916 | 81 | ||
| AK153085.1 | - | 7 - 8 | 936 | 81 | ||
| AK162148.1 | - | 7 - 8 | 912 | 81 | ||
| AK169254.1 | - | 7 - 8 | 912 | 81 | ||
| AK189959.1 | - | 2 - 3 | 197 | 81 | ||
| AK190473.1 | - | 2 - 3 | 290 | 81 | ||
| AK202419.1 | - | 2 - 3 | 291 | 81 | ||
| AK214998.1 | - | 2 - 3 | 228 | 81 | ||
| AK217952.1 | - | 2 - 3 | 290 | 81 | ||
| BC011301.1 | - | 7 - 8 | 889 | 81 | ||
| BC031849.1 | - | 7 - 8 | 937 | 5067510 | 81 | |
| DQ402975.1 | - | 5 - 6 | 723 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sdha
Genomic Map