Assay Details
| Gene Symbol: Sdha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023281.1 | NP_075770.1 | 8 - 9 | 1098 | 73 | |
| GenBank mRNA | AF095938.1 | - | 6 - 7 | 847 | 73 | |
| AK010561.1 | - | 8 - 9 | 1087 | 73 | ||
| AK029520.1 | - | 8 - 9 | 1098 | 73 | ||
| AK034928.1 | - | 8 - 9 | 1099 | 73 | ||
| AK049590.1 | - | 8 - 9 | 1085 | 73 | ||
| AK050475.1 | - | 8 - 9 | 1085 | 73 | ||
| AK075990.1 | - | 8 - 9 | 1082 | 73 | ||
| AK145923.1 | - | 8 - 9 | 1096 | 73 | ||
| AK147286.1 | - | 8 - 9 | 1085 | 73 | ||
| AK147624.1 | - | 8 - 9 | 1085 | 73 | ||
| AK153085.1 | - | 8 - 9 | 1105 | 73 | ||
| AK162148.1 | - | 8 - 9 | 1081 | 73 | ||
| AK169254.1 | - | 8 - 9 | 1081 | 73 | ||
| BC011301.1 | - | 8 - 9 | 1058 | 73 | ||
| BC031849.1 | - | 8 - 9 | 1106 | 5067510 | 73 | |
| DQ402975.1 | - | 6 - 7 | 892 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sdha
Genomic Map