Assay Details
| Gene Symbol: Sdha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023281.1 | NP_075770.1 | 9 - 10 | 1292 | 73 | |
| GenBank mRNA | AF095938.1 | - | 7 - 8 | 1041 | 73 | |
| AK010561.1 | - | 9 - 10 | 1281 | 73 | ||
| AK029520.1 | - | 9 - 10 | 1292 | 73 | ||
| AK034928.1 | - | 9 - 10 | 1293 | 73 | ||
| AK049590.1 | - | 9 - 10 | 1279 | 73 | ||
| AK050475.1 | - | 9 - 10 | 1279 | 73 | ||
| AK075990.1 | - | 9 - 10 | 1276 | 73 | ||
| AK147286.1 | - | 9 - 10 | 1279 | 73 | ||
| AK147624.1 | - | 9 - 10 | 1279 | 73 | ||
| AK153085.1 | - | 9 - 10 | 1299 | 73 | ||
| AK162148.1 | - | 9 - 10 | 1275 | 73 | ||
| AK169254.1 | - | 9 - 10 | 1275 | 73 | ||
| AK186810.1 | - | 1 - 2 | 126 | 73 | ||
| AK207174.1 | - | 1 - 2 | 126 | 73 | ||
| AK216523.1 | - | 1 - 2 | 118 | 73 | ||
| BC011301.1 | - | 9 - 10 | 1252 | 73 | ||
| BC031849.1 | - | 9 - 10 | 1300 | 5067510 | 73 | |
| DQ402975.1 | - | 7 - 8 | 1086 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sdha
Genomic Map