Assay Details
| Gene Symbol: Zdhhc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027704.2 | NP_081980.1 | 3 - 4 | 468 | 58 | |
| GenBank mRNA | AK016865.1 | - | 3 - 4 | 479 | 58 | |
| AY668948.1 | - | 2 - 3 | 409 | 58 | ||
| BC116803.1 | - | 3 - 4 | 468 | 40126122 | 58 | |
| BC116807.1 | - | 3 - 4 | 468 | 40126126 | 58 | |
| BC144832.1 | - | 3 - 4 | 468 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc11
Genomic Map