Assay Details
| Gene Symbol: Zdhhc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027704.2 | NP_081980.1 | 5 - 6 | 694 | 81 | |
| GenBank mRNA | AK016865.1 | - | 5 - 6 | 705 | 81 | |
| AY668948.1 | - | 4 - 5 | 635 | 81 | ||
| BC116803.1 | - | 5 - 6 | 694 | 40126122 | 81 | |
| BC116807.1 | - | 5 - 6 | 694 | 40126126 | 81 | |
| BC144831.1 | - | 5 - 6 | 686 | 81 | ||
| BC144832.1 | - | 5 - 6 | 694 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc11
Genomic Map