Assay Details
| Gene Symbol: Zdhhc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016865.1 | - | 7 - 8 | 1026 | 68 | |
| BC144832.1 | - | 7 - 8 | 1015 | 68 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zdhhc11| Gene Symbol: Zdhhc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016865.1 | - | 7 - 8 | 1026 | 68 | |
| BC144832.1 | - | 7 - 8 | 1015 | 68 | ||
Set Membership: |
|