Assay Details
| Gene Symbol: Exoc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177333.3 | NP_796307.2 | 4 - 5 | 1136 | 87 | |
| GenBank mRNA | AK088333.1 | - | 4 - 5 | 1130 | 87 | |
| AK089979.1 | - | 4 - 5 | 1146 | 87 | ||
| AK131141.1 | - | 4 - 5 | 1120 | 87 | ||
| AK141537.1 | - | 4 - 5 | 1136 | 87 | ||
| BC031743.1 | - | 4 - 5 | 1124 | 4187527 | 87 | |
More Information
Set Membership: |
|

Exoc3
Genomic Map